Product Details

SNP ID
rs963328
Assay Type
Validated
NCBI dbSNP Submissions
52
Location
Chr.1:212864319 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
ACAATTAACTGGATCTGAAAATGTT[G/T]ACTAGTGAATGGTCCTGGGCAGCAG
Phenotype
MIM: 609144 MIM: 610864
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
FLVCR1 PubMed Links
Additional Information
For this assay, SNP(s) [rs41258005] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
FLVCR1
Gene Name
feline leukemia virus subgroup C cellular receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014053.3 Intron NP_054772.1
XM_011509446.2 Intron XP_011507748.1
XM_011509447.2 Intron XP_011507749.1
XM_011509448.1 Intron XP_011507750.1
Gene
FLVCR1-AS1
Gene Name
FLVCR1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details