Product Details

SNP ID
rs1201525
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:5237824 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGCCCGCCTGAGGGAGCCCTCCGG[A/G]AGTGAGCGCACGCAACCGTGCAGTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00526 PubMed Links

Gene Details

Gene
LINC00526
Gene Name
long intergenic non-protein coding RNA 526
There are no transcripts associated with this gene.

Gene
LINC00667
Gene Name
long intergenic non-protein coding RNA 667
There are no transcripts associated with this gene.

View Full Product Details