Product Details

SNP ID
rs1498363
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:214349657 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTTCCACTGAAGTATCCTCAAG[C/T]ACATTAATTAGCTATTGAAGGAACA
Phenotype
MIM: 603155
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTPN14 PubMed Links

Gene Details

Gene
PTPN14
Gene Name
protein tyrosine phosphatase, non-receptor type 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005401.4 12482 UTR 3 NP_005392.2
XM_017001941.1 12482 Intron XP_016857430.1

View Full Product Details