Product Details

SNP ID
rs437701
Assay Type
Functionally tested
NCBI dbSNP Submissions
31
Location
Chr.1:212295668 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGGCCAAAGAAGACAGGAGAGAG[C/T]GGAGACAAGGCTGTCAGGGGAAGAG
Phenotype
MIM: 601643
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP2R5A PubMed Links

Gene Details

Gene
PPP2R5A
Gene Name
protein phosphatase 2 regulatory subunit B'alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199756.1 Intron NP_001186685.1
NM_006243.3 Intron NP_006234.1

View Full Product Details