Product Details

SNP ID
rs1058151
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:649759 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGGTTTCACATTATTTTGCTACA[C/T]ACAGTACCAATTTTTCCAAAAGCCA
Phenotype
MIM: 616990 MIM: 188350
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLUL1 PubMed Links

Gene Details

Gene
CLUL1
Gene Name
clusterin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289036.1 705 Intron NP_001275965.1
NM_001318522.1 705 Intron NP_001305451.1
NM_014410.4 705 Intron NP_055225.1
NM_199167.1 705 Intron NP_954636.1
XM_005258103.3 705 Intron XP_005258160.1
XM_006722322.2 705 Intron XP_006722385.1
XM_011525648.2 705 Intron XP_011523950.1
XM_011525649.1 705 Intron XP_011523951.1
XM_011525651.2 705 Intron XP_011523953.1
XM_011525652.1 705 Intron XP_011523954.1
XM_011525653.2 705 Intron XP_011523955.1
XM_017025707.1 705 Intron XP_016881196.1
Gene
TYMS
Gene Name
thymidylate synthetase
There are no transcripts associated with this gene.

Gene
TYMSOS
Gene Name
TYMS opposite strand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012716.2 705 UTR 3 NP_001012734.2

View Full Product Details