Product Details

SNP ID
rs3755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:26455824 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGTTCCCGCCGAAGGACGTATTC[A/G]CTGAACTGGGACGAGTCTACTCCTC
Phenotype
MIM: 613420
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCTD1 PubMed Links

Gene Details

Gene
KCTD1
Gene Name
potassium channel tetramerization domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136205.2 833 Silent Mutation AGC,AGT S231S NP_001129677.1
NM_001142730.2 833 Silent Mutation AGC,AGT S839S NP_001136202.1
NM_001258221.1 833 Silent Mutation AGC,AGT S231S NP_001245150.1
NM_001258222.1 833 Silent Mutation AGC,AGT S239S NP_001245151.1
NM_198991.3 833 Silent Mutation AGC,AGT S231S NP_945342.1
XM_017025708.1 833 Silent Mutation AGC,AGT S231S XP_016881197.1

View Full Product Details