Product Details

SNP ID
rs1378543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:114521576 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGAAGGGTTCTTAGACATCACAGT[C/T]AGGGTGCTCGGAGTGATTATTGCTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NXPE1 PubMed Links

Gene Details

Gene
NXPE1
Gene Name
neurexophilin and PC-esterase domain family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152315.3 2763 Intron NP_689528.2
XM_011542594.2 2763 UTR 3 XP_011540896.1
XM_011542595.2 2763 UTR 3 XP_011540897.1
XM_011542596.2 2763 UTR 3 XP_011540898.1
XM_011542597.2 2763 UTR 3 XP_011540899.1
XM_011542598.2 2763 UTR 3 XP_011540900.1
XM_011542599.2 2763 UTR 3 XP_011540901.1
Gene
NXPE2
Gene Name
neurexophilin and PC-esterase domain family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182495.5 2763 Intron NP_872301.2
XM_011542604.2 2763 Intron XP_011540906.1
XM_017017206.1 2763 Intron XP_016872695.1
XM_017017207.1 2763 Intron XP_016872696.1
XM_017017208.1 2763 Intron XP_016872697.1
XM_017017209.1 2763 Intron XP_016872698.1
XM_017017210.1 2763 Intron XP_016872699.1
XM_017017211.1 2763 Intron XP_016872700.1
XM_017017212.1 2763 Intron XP_016872701.1

View Full Product Details