Product Details

SNP ID
rs1456888
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:113976818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTACCTGCCAGCTCCATCTCCC[C/T]TTTGCTGCCCGTATGCTGGCCCTCT
Phenotype
MIM: 182139
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HTR3A PubMed Links

Gene Details

Gene
HTR3A
Gene Name
5-hydroxytryptamine receptor 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000869.5 Intron NP_000860.2
NM_001161772.2 Intron NP_001155244.1
NM_213621.3 Intron NP_998786.2

View Full Product Details