Product Details

SNP ID
rs1052369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:23599180 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGTACAGTCTTTGGTAGGTGATGA[A/T]TCATTTTCCCTGCTATGGGTAATCT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD29 PubMed Links

Gene Details

Gene
ANKRD29
Gene Name
ankyrin repeat domain 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308238.1 2931 UTR 3 NP_001295167.1
NM_173505.3 2931 UTR 3 NP_775776.2
XM_011525830.2 2931 UTR 3 XP_011524132.1
XM_011525831.2 2931 UTR 3 XP_011524133.1
XM_011525832.2 2931 UTR 3 XP_011524134.1
XM_017025561.1 2931 UTR 3 XP_016881050.1

View Full Product Details