Product Details

SNP ID
rs1049461
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102349840 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGAATGTTTTAGCTATCAAACAG[C/T]ACTGTCACCTACTCATGCACAAAAC
Phenotype
MIM: 601712
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BIRC2 PubMed Links

Gene Details

Gene
BIRC2
Gene Name
baculoviral IAP repeat containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166.4 1438 UTR 5 NP_001157.1
NM_001256163.1 1438 UTR 5 NP_001243092.1
NM_001256166.1 1438 Intron NP_001243095.1

View Full Product Details