Product Details

SNP ID
rs1465592
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17515413 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGAGCAGACCAGGGCCCCTGGGG[C/G]GCCTCTCAGTGGGATAGGGCTGCAG
Phenotype
MIM: 609967 MIM: 604951 MIM: 600691
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
FAM129C PubMed Links
Additional Information
For this assay, SNP(s) [rs77487659] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
FAM129C
Gene Name
family with sequence similarity 129 member C
There are no transcripts associated with this gene.

Gene
PGLS
Gene Name
6-phosphogluconolactonase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012088.2 Intron NP_036220.1
XM_011527897.2 Intron XP_011526199.1
Gene
SLC27A1
Gene Name
solute carrier family 27 member 1
There are no transcripts associated with this gene.

View Full Product Details