Product Details

SNP ID
rs1061929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134605403 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGAATCTCAAACATATAAAAAAA[A/G]AATTCAGTTTCTTAGGAAATTATAA
Phenotype
MIM: 607690
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SAR1B PubMed Links

Gene Details

Gene
SAR1B
Gene Name
secretion associated Ras related GTPase 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033503.2 2278 UTR 3 NP_001028675.1
NM_016103.3 2278 UTR 3 NP_057187.1
XM_017009521.1 2278 UTR 3 XP_016865010.1

View Full Product Details