Product Details

SNP ID
rs15978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:7285056 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATGGAAGAATGGCAAATGAAAA[C/T]TGGCCCACATTTTCTTGTTCCTTCT
Phenotype
MIM: 600868
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SSR1 PubMed Links

Gene Details

Gene
SSR1
Gene Name
signal sequence receptor subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292008.1 5863 UTR 3 NP_001278937.1
NM_003144.4 5863 UTR 3 NP_003135.2

View Full Product Details