Product Details

SNP ID
rs1285875
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:7115694 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGTTCTGGTGCTTTTCTGTCTTCTC[C/G]TCTAACCTATAAACTGCCAACTCCC
Phenotype
MIM: 602209
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RREB1 PubMed Links

Gene Details

Gene
RREB1
Gene Name
ras responsive element binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003698.3 Intron NP_001003698.1
NM_001003699.3 Intron NP_001003699.1
NM_001003700.1 Intron NP_001003700.1
NM_001168344.1 Intron NP_001161816.1
XM_006715157.3 Intron XP_006715220.1
XM_011514794.2 Intron XP_011513096.1
XM_011514795.2 Intron XP_011513097.1

View Full Product Details