Product Details

SNP ID
rs1678822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:21296056 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCAGACAAGAAGAGAATCTTGGGA[G/T]CTGCAAGATAAAGTAATGTATCATT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF708 PubMed Links

Gene Details

Gene
ZNF708
Gene Name
zinc finger protein 708
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297560.1 Intron NP_001284489.1
NM_001297561.1 Intron NP_001284490.1
NM_021269.2 Intron NP_067092.2
XM_017027201.1 Intron XP_016882690.1
XM_017027202.1 Intron XP_016882691.1
XM_017027203.1 Intron XP_016882692.1
XM_017027204.1 Intron XP_016882693.1

View Full Product Details