Product Details

SNP ID
rs904643
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:124301507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCTCCAGTGCTCGGCCTCCCTG[C/T]GGGGCACGGTACATGGACTGTGGAC
Phenotype
MIM: 615927
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FAM101A PubMed Links
Additional Information
For this assay, SNP(s) [rs77263762] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
FAM101A
Gene Name
family with sequence similarity 101 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181709.4 Intron NP_859060.3
Gene
LOC105370045
Gene Name
filamin-interacting protein FAM101A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011539088.1 Intron XP_011537390.1
Gene
ZNF664-FAM101A
Gene Name
filamin-interacting protein FAM101A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204299.1 Intron NP_001191228.1

View Full Product Details