Product Details

SNP ID
rs923996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:23209881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTAGAGTATTCAGTTTCCCTTTC[A/G]TGGGGGTGTATTGCTGATCTCTGTT
Phenotype
MIM: 602163
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBE2E2 PubMed Links

Gene Details

Gene
UBE2E2
Gene Name
ubiquitin conjugating enzyme E2 E2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152653.3 Intron NP_689866.1
XM_005265432.2 Intron XP_005265489.1
XM_005265433.4 Intron XP_005265490.1
XM_011534074.2 Intron XP_011532376.1
XM_011534075.1 Intron XP_011532377.1
XM_011534076.2 Intron XP_011532378.1
XM_017007126.1 Intron XP_016862615.1
Gene
UBE2E2-AS1
Gene Name
UBE2E2 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details