Product Details

SNP ID
rs1044939
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:57055922 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCAATTCCACAAACATGGAAAATTA[C/T]GGTAACTGCAGGCTGTAACAGTTTC
Phenotype
MIM: 601478 MIM: 616496
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYO1A PubMed Links

Gene Details

Gene
MYO1A
Gene Name
myosin IA
There are no transcripts associated with this gene.

Gene
NEMP1
Gene Name
nuclear envelope integral membrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130963.1 5338 UTR 3 NP_001124435.1
NM_015257.2 5338 UTR 3 NP_056072.2
XM_011538056.2 5338 UTR 3 XP_011536358.1
XM_011538057.2 5338 UTR 3 XP_011536359.1
XM_011538058.2 5338 UTR 3 XP_011536360.1
XM_011538059.1 5338 UTR 3 XP_011536361.1
XM_011538060.2 5338 Intron XP_011536362.1
XM_017019077.1 5338 UTR 3 XP_016874566.1
XM_017019078.1 5338 UTR 3 XP_016874567.1
XM_017019079.1 5338 UTR 3 XP_016874568.1

View Full Product Details