Product Details

SNP ID
rs590580
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118195389 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGTCATGTGTCCTCAGGGCTGGT[A/G]CGATAGGAACAAACAAAGCCCATCA
Phenotype
MIM: 609770
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
JAML PubMed Links

Gene Details

Gene
JAML
Gene Name
junction adhesion molecule like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098526.1 Intron NP_001091996.1
NM_001286570.1 Intron NP_001273499.1
NM_001286571.1 Intron NP_001273500.1
NM_153206.2 Intron NP_694938.2
XM_005271407.3 Intron XP_005271464.1
XM_017017213.1 Intron XP_016872702.1
XM_017017214.1 Intron XP_016872703.1
XM_017017215.1 Intron XP_016872704.1

View Full Product Details