Product Details

SNP ID
rs14189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:47655650 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATACTCTCCTGTTTGGAGACAGAGG[A/T]AGAACCAGGTCAGTCTGTCTCTTTT
Phenotype
MIM: 601937 MIM: 610013
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NCOA3 PubMed Links

Gene Details

Gene
NCOA3
Gene Name
nuclear receptor coactivator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174087.1 6769 UTR 3 NP_001167558.1
NM_001174088.1 6769 UTR 3 NP_001167559.1
NM_006534.3 6769 UTR 3 NP_006525.2
NM_181659.2 6769 UTR 3 NP_858045.1
Gene
SULF2
Gene Name
sulfatase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161841.1 6769 Intron NP_001155313.1
NM_018837.3 6769 Intron NP_061325.1
NM_198596.2 6769 Intron NP_940998.2
XM_005260458.2 6769 UTR 3 XP_005260515.1
XM_006723827.1 6769 Intron XP_006723890.1
XM_006723828.1 6769 Intron XP_006723891.1
XM_006723829.2 6769 Intron XP_006723892.1
XM_006723830.1 6769 Intron XP_006723893.1
XM_011528913.1 6769 Intron XP_011527215.1
XM_011528914.1 6769 Intron XP_011527216.1
XM_011528915.1 6769 Intron XP_011527217.1
XM_011528916.1 6769 Intron XP_011527218.1
XM_017027955.1 6769 UTR 3 XP_016883444.1

View Full Product Details