Product Details

SNP ID
rs7759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119096581 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTGCTTAGTTCTTACTAATCAGA[A/G]CACTTAATTTACAATCAAATGACAT
Phenotype
MIM: 191350 MIM: 601772 MIM: 609806
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPAGT1 PubMed Links
Additional Information
For this assay, SNP(s) [rs28990975] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
DPAGT1
Gene Name
dolichyl-phosphate N-acetylglucosaminephosphotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001382.3 2282 UTR 3 NP_001373.2
XM_005271422.2 2282 UTR 3 XP_005271479.1
XM_011542648.1 2282 UTR 3 XP_011540950.1
XM_017017293.1 2282 UTR 3 XP_016872782.1
XM_017017294.1 2282 UTR 3 XP_016872783.1
XM_017017295.1 2282 UTR 3 XP_016872784.1
Gene
H2AFX
Gene Name
H2A histone family member X
There are no transcripts associated with this gene.

Gene
HMBS
Gene Name
hydroxymethylbilane synthase
There are no transcripts associated with this gene.

View Full Product Details