Product Details

SNP ID
rs1020052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96186333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTCAACTGGCCAACTATACTTTC[C/G]AGGTGGACCTGGAGAACTGAAGGAA
Phenotype
MIM: 616712
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
STARD7 PubMed Links

Gene Details

Gene
STARD7
Gene Name
StAR related lipid transfer domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020151.3 1911 UTR 3 NP_064536.2
XM_011511499.1 1911 UTR 3 XP_011509801.1

View Full Product Details