Product Details

SNP ID
rs1724119
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96124109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCTTTTTCAGGGCAGGGGACTCC[A/C]ACCCATGTGGGCTCTCCCCAGGCCC
Phenotype
MIM: 104260 MIM: 608860
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADRA2B PubMed Links

Gene Details

Gene
ADRA2B
Gene Name
adrenoceptor alpha 2B
There are no transcripts associated with this gene.

Gene
ASTL
Gene Name
astacin-like metallo-endopeptidase (M12 family)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002036.3 1852 Missense Mutation TGG,TTG W346L NP_001002036.3
XM_011511205.2 1852 Missense Mutation TGG,TTG W351L XP_011509507.1
XM_011511206.2 1852 Missense Mutation TGG,TTG W347L XP_011509508.1
XM_011511207.2 1852 Missense Mutation TGG,TTG W333L XP_011509509.1
XM_011511208.2 1852 Intron XP_011509510.1

View Full Product Details