Product Details

SNP ID
rs1043797
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:215071 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATACCACATACCACACATAGCC[C/A]CAGAAACATCATCTTGAAATAAAGA
Phenotype
MIM: 615417 MIM: 609146 MIM: 604481
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
BET1L PubMed Links

Gene Details

Gene
BET1L
Gene Name
Bet1 golgi vesicular membrane trafficking protein like
There are no transcripts associated with this gene.

Gene
MIR6743
Gene Name
microRNA 6743
There are no transcripts associated with this gene.

Gene
RIC8A
Gene Name
RIC8 guanine nucleotide exchange factor A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286134.1 2847 UTR 3 NP_001273063.1
NM_021932.5 2847 UTR 3 NP_068751.4
XM_005253052.3 2847 UTR 3 XP_005253109.1
XM_017018107.1 2847 UTR 3 XP_016873596.1
Gene
SIRT3
Gene Name
sirtuin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017524.2 2847 UTR 3 NP_001017524.1
NM_012239.5 2847 UTR 3 NP_036371.1
XM_005252835.1 2847 Intron XP_005252892.1
XM_011519956.1 2847 Intron XP_011518258.1
XM_011519957.1 2847 Intron XP_011518259.1
XM_017017428.1 2847 Intron XP_016872917.1
XM_017017429.1 2847 Intron XP_016872918.1
XM_017017430.1 2847 Intron XP_016872919.1
XM_017017431.1 2847 Intron XP_016872920.1

View Full Product Details