Product Details
- SNP ID
-
rs3850191
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:171843911 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AACCTTGAAAAGTAATTTTAAAATA[C/G]AGAACAAAGTAAAATAATTTTCAAA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM212
PubMed Links
Gene Details
- Gene
- TMEM212
- Gene Name
- transmembrane protein 212
View Full Product Details