Product Details
- SNP ID
-
rs1353776
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:134358628 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGTGGAGGTGCTGTCTGGGGGGCC[C/G]TCAGTCTGGGTGCTCCCATCTCTGC
- Phenotype
-
MIM: 614658
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
AMOTL2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117533420] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AMOTL2
- Gene Name
- angiomotin like 2
- Gene
- MIR6827
- Gene Name
- microRNA 6827
There are no transcripts associated with this gene.
- Gene
- RPL39P5
- Gene Name
- ribosomal protein L39 pseudogene 5
There are no transcripts associated with this gene.
View Full Product Details