Product Details

SNP ID
rs1051043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:100333857 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGAGTGCTCCAGAGGAGACCCA[A/G]ATACATCAACCAAGGACTTCCCTGA
Phenotype
MIM: 609911 MIM: 191050
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC25A47 PubMed Links

Gene Details

Gene
SLC25A47
Gene Name
solute carrier family 25 member 47
There are no transcripts associated with this gene.

Gene
WARS
Gene Name
tryptophanyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004184.3 2789 UTR 3 NP_004175.2
NM_173701.1 2789 UTR 3 NP_776049.1
NM_213645.1 2789 UTR 3 NP_998810.1
NM_213646.1 2789 UTR 3 NP_998811.1
XM_005268044.3 2789 UTR 3 XP_005268101.1
XM_006720249.3 2789 UTR 3 XP_006720312.1
XM_011537133.2 2789 UTR 3 XP_011535435.1
XM_011537135.2 2789 UTR 3 XP_011535437.1
XM_011537136.2 2789 UTR 3 XP_011535438.1
XM_017021627.1 2789 UTR 3 XP_016877116.1
XM_017021628.1 2789 UTR 3 XP_016877117.1
XM_017021629.1 2789 UTR 3 XP_016877118.1

View Full Product Details