Product Details

SNP ID
rs1501743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:38847891 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGGTCTTGGGTGTTTCTTCCGCA[A/G]GGACCCGAGCCGCATTGGGGAGAAC
Phenotype
MIM: 601743
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OSMR PubMed Links

Gene Details

Gene
OSMR
Gene Name
oncostatin M receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168355.2 Intron NP_001161827.1
NM_001323504.1 Intron NP_001310433.1
NM_001323505.1 Intron NP_001310434.1
NM_001323506.1 Intron NP_001310435.1
NM_001323507.1 Intron NP_001310436.1
NM_003999.2 Intron NP_003990.1
XM_005248384.1 Intron XP_005248441.1
XM_005248386.2 Intron XP_005248443.1
XM_005248387.2 Intron XP_005248444.1
XM_011514161.2 Intron XP_011512463.1
XM_017010019.1 Intron XP_016865508.1
XM_017010020.1 Intron XP_016865509.1
Gene
OSMR-AS1
Gene Name
OSMR antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details