Product Details
- SNP ID
-
rs1504974
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:119269670 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATTTTCGAAAACACAGAAGCTCTA[A/T]GTAGAAAAAGCAGTATCCCTAGAGG
- Phenotype
-
MIM: 612111
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TNFAIP8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77856693] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TNFAIP8
- Gene Name
- TNF alpha induced protein 8
View Full Product Details