Product Details

SNP ID
rs3867247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124408234 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTTTAGGTGAACAATGCTTATTT[A/G]TCCTCTTCCTTTTCAATCCATTGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR8B3 PubMed Links

Gene Details

Gene
OR8B3
Gene Name
olfactory receptor family 8 subfamily B member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005467.1 Intron NP_001005467.1
XM_017017716.1 Intron XP_016873205.1
XM_017017717.1 Intron XP_016873206.1
XM_017017718.1 Intron XP_016873207.1

View Full Product Details