Product Details

SNP ID
rs3915491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62213591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAATAAATGGAGAGGGGATGCT[C/G]GCTTTGCTTCCACTGCAAATCCTCC
Phenotype
MIM: 604398
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SCGB2A1 PubMed Links

Gene Details

Gene
SCGB2A1
Gene Name
secretoglobin family 2A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002407.2 Intron NP_002398.1

View Full Product Details