Product Details

SNP ID
rs10092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:73075026 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATCTGTCTACCATAAATTGTCTTC[A/G]TGACAAATGCAGAATTTCTTTTATT
Phenotype
MIM: 615929 MIM: 610428
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD17 PubMed Links

Gene Details

Gene
ANKRD17
Gene Name
ankyrin repeat domain 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286771.2 8394 UTR 3 NP_001273700.1
NM_015574.1 8394 UTR 3 NP_056389.1
NM_032217.4 8394 UTR 3 NP_115593.3
NM_198889.2 8394 UTR 3 NP_942592.1
XM_005265671.3 8394 UTR 3 XP_005265728.1
XM_005265672.3 8394 Intron XP_005265729.1
XM_005265673.3 8394 Intron XP_005265730.1
XM_017008011.1 8394 UTR 3 XP_016863500.1
XM_017008012.1 8394 UTR 3 XP_016863501.1
XM_017008013.1 8394 UTR 3 XP_016863502.1
XM_017008014.1 8394 Intron XP_016863503.1
XM_017008015.1 8394 Intron XP_016863504.1
XM_017008016.1 8394 Intron XP_016863505.1
XM_017008017.1 8394 Intron XP_016863506.1
Gene
COX18
Gene Name
COX18, cytochrome c oxidase assembly factor
There are no transcripts associated with this gene.

View Full Product Details