Product Details

SNP ID
rs1054489
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:151101146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGATGTTTTTGGATCTGACATA[C/G]CCCAAACCAGGGCAGAGGCTGTAGC
Phenotype
MIM: 114070
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ANXA6 PubMed Links

Gene Details

Gene
ANXA6
Gene Name
annexin A6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001155.4 2534 UTR 3 NP_001146.2
NM_001193544.1 2534 UTR 3 NP_001180473.1
XM_005268432.4 2534 UTR 3 XP_005268489.1

View Full Product Details