Product Details

SNP ID
hCV7515550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32116563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGATGGCTGGGAGCAGCAGGTGG[C/T]CCTGGGGAACAGATGGGCCAGGCCA
Phenotype
MIM: 600984 MIM: 600985
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATF6B PubMed Links

Gene Details

Gene
ATF6B
Gene Name
activating transcription factor 6 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136153.1 1832 Missense Mutation GAC,GGC D597G NP_001129625.1
NM_004381.4 1832 Missense Mutation GAC,GGC D600G NP_004372.3
Gene
TNXB
Gene Name
tenascin XB
There are no transcripts associated with this gene.

View Full Product Details