Product Details
- SNP ID
-
rs940775
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:149433759 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGAATCTGGATCAGGTGCAGGCAG[A/T]GCATGCCCCACCCGGGGATGCCCCC
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF777
PubMed Links
Gene Details
- Gene
- ZNF777
- Gene Name
- zinc finger protein 777
View Full Product Details