Product Details

SNP ID
rs1202432
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149139420 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTATTCTAAATTCCAAATGTAACTT[A/G]AAGTATTAAATCTAATGATGTGCCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF398 PubMed Links

Gene Details

Gene
ZNF398
Gene Name
zinc finger protein 398
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020781.3 Intron NP_065832.1
NM_170686.2 Intron NP_733787.1
XM_011516439.2 Intron XP_011514741.1
XM_011516440.2 Intron XP_011514742.1
XM_011516441.2 Intron XP_011514743.1

View Full Product Details