Product Details

SNP ID
rs1060053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89644477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTATTTACAACAGCATTCTAGGA[A/G]AAAGGTGAATGGGCCCCTCTCCTCA
Phenotype
MIM: 164010 MIM: 179780
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHMP1A PubMed Links

Gene Details

Gene
CHMP1A
Gene Name
charged multivesicular body protein 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083314.3 2377 UTR 3 NP_001076783.1
NM_002768.4 2377 UTR 3 NP_002759.2
XM_011523098.1 2377 Intron XP_011521400.1
Gene
DPEP1
Gene Name
dipeptidase 1 (renal)
There are no transcripts associated with this gene.

View Full Product Details