Product Details

SNP ID
rs847163
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:176129895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTTTTTTCTTTTCCCTCCAGAGC[C/T]GGGGTTTGTAAACCGAGGCCAGAGT
Phenotype
MIM: 142984 MIM: 142980 MIM: 142985 MIM: 142982
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HOXD-AS2 PubMed Links

Gene Details

Gene
HOXD-AS2
Gene Name
HOXD cluster antisense RNA 2
There are no transcripts associated with this gene.

Gene
HOXD10
Gene Name
homeobox D10
There are no transcripts associated with this gene.

Gene
HOXD3
Gene Name
homeobox D3
There are no transcripts associated with this gene.

Gene
HOXD8
Gene Name
homeobox D8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199746.1 156 UTR 5 NP_001186675.1
NM_001199747.1 156 Intron NP_001186676.1
NM_019558.3 156 UTR 5 NP_062458.1
Gene
HOXD9
Gene Name
homeobox D9
There are no transcripts associated with this gene.

Gene
LOC100129455
Gene Name
uncharacterized LOC100129455
There are no transcripts associated with this gene.

View Full Product Details