Product Details
- SNP ID
-
rs921397
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:37848972 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAGGTCACCGCTCAAACGGCCACAA[C/T]GAGGGAGCCGATTTAGATCCTCTGG
- Phenotype
-
MIM: 607421
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NME8
PubMed Links
Gene Details
- Gene
- NME8
- Gene Name
- NME/NM23 family member 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016616.4 |
281 |
UTR 5 |
|
|
NP_057700.3 |
View Full Product Details