Product Details

SNP ID
rs9252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42402831 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTCTCGGGGCCTCCCTTCAGGCT[A/G]TGACTGGGGAGAGGGGCTGTTCTTG
Phenotype
MIM: 603198
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTRF PubMed Links

Gene Details

Gene
PTRF
Gene Name
polymerase I and transcript release factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012232.5 3252 UTR 3 NP_036364.2
XM_005257242.3 3252 Intron XP_005257299.1

View Full Product Details