Product Details

SNP ID
rs1804196
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:112878589 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAAAAACGTGGACACTGCCCACT[G/T]CATTAAGTTTAAAGTGCTCCCTATA
Phenotype
MIM: 125265 MIM: 182175
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
REEP5 PubMed Links

Gene Details

Gene
REEP5
Gene Name
receptor accessory protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005669.4 911 UTR 3 NP_005660.4
XM_017009844.1 911 Intron XP_016865333.1
Gene
SRP19
Gene Name
signal recognition particle 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204193.1 911 Intron NP_001191122.1
NM_001204194.1 911 Intron NP_001191123.1
NM_001204196.1 911 Intron NP_001191125.1
NM_001204199.1 911 Intron NP_001191128.1
NM_003135.2 911 Intron NP_003126.1

View Full Product Details