Product Details

SNP ID
hCV7531339
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:125003422 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATCATGTTCTCAAAGACCTTGGT[C/T]TTGGAGGTAGGTTCATATTGATTTT
Phenotype
MIM: 602019
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105375744 PubMed Links

Gene Details

Gene
LOC105375744
Gene Name
uncharacterized LOC105375744
There are no transcripts associated with this gene.

Gene
SQLE
Gene Name
squalene epoxidase
There are no transcripts associated with this gene.

View Full Product Details