Product Details

SNP ID
rs1048654
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22972170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGATCTAGCTTTAGGTGAACTAG[C/T]TTCAAGCAAACTGACTAGTATCAAA
Phenotype
MIM: 609066
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AJUBA PubMed Links

Gene Details

Gene
AJUBA
Gene Name
ajuba LIM protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289097.1 3266 Intron NP_001276026.1
NM_032876.5 3266 UTR 3 NP_116265.1
NM_198086.2 3266 UTR 3 NP_932352.1

View Full Product Details