Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001130706.2 | 1905 | UTR 3 | NP_001124178.1 | ||
NM_001130708.2 | 1905 | UTR 3 | NP_001124180.1 | ||
NM_001282968.1 | 1905 | UTR 3 | NP_001269897.1 | ||
NM_001282969.1 | 1905 | UTR 3 | NP_001269898.1 | ||
NM_001282970.1 | 1905 | UTR 3 | NP_001269899.1 | ||
NM_021944.3 | 1905 | UTR 3 | NP_068763.2 | ||
XM_005267971.1 | 1905 | UTR 3 | XP_005268028.1 | ||
XM_005267972.1 | 1905 | UTR 3 | XP_005268029.1 | ||
XM_006720231.1 | 1905 | UTR 3 | XP_006720294.1 | ||
XM_006720232.2 | 1905 | UTR 3 | XP_006720295.1 | ||
XM_011537059.1 | 1905 | UTR 3 | XP_011535361.1 | ||
XM_011537060.2 | 1905 | UTR 3 | XP_011535362.1 | ||
XM_011537062.1 | 1905 | UTR 3 | XP_011535364.1 | ||
XM_017021569.1 | 1905 | UTR 3 | XP_016877058.1 | ||
XM_017021570.1 | 1905 | UTR 3 | XP_016877059.1 | ||
XM_017021571.1 | 1905 | UTR 3 | XP_016877060.1 | ||
XM_017021572.1 | 1905 | UTR 3 | XP_016877061.1 | ||
XM_017021573.1 | 1905 | UTR 3 | XP_016877062.1 | ||
XM_017021574.1 | 1905 | UTR 3 | XP_016877063.1 | ||
XM_017021575.1 | 1905 | UTR 3 | XP_016877064.1 | ||
XM_017021576.1 | 1905 | UTR 3 | XP_016877065.1 | ||
XM_017021577.1 | 1905 | UTR 3 | XP_016877066.1 |