Product Details

SNP ID
rs935901
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.15:43359058 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTTTTCCCATTCCTCAACCCACCCA[C/G]CACTCAATTTTGCAACTAGTTATGG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADAL PubMed Links

Gene Details

Gene
ADAL
Gene Name
adenosine deaminase like
There are no transcripts associated with this gene.

Gene
ZSCAN29
Gene Name
zinc finger and SCAN domain containing 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152455.3 5161 UTR 3 NP_689668.3
XM_006720401.3 5161 UTR 3 XP_006720464.1
XM_011521266.2 5161 UTR 3 XP_011519568.1
XM_017021949.1 5161 Intron XP_016877438.1

View Full Product Details