Product Details

SNP ID
rs1412493
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:31807875 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTGTTAAAAGAGAAAATAAGAGA[C/G]AGGGAAAATTCTTCAAATGGTATTA
Phenotype
MIM: 610577
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP12 PubMed Links

Gene Details

Gene
ARHGAP12
Gene Name
Rho GTPase activating protein 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270695.1 Intron NP_001257624.1
NM_001270696.1 Intron NP_001257625.1
NM_001270697.1 Intron NP_001257626.1
NM_001270698.1 Intron NP_001257627.1
NM_001270699.1 Intron NP_001257628.1
NM_018287.6 Intron NP_060757.4
XM_005252644.1 Intron XP_005252701.1
XM_011519761.1 Intron XP_011518063.1
XM_017016954.1 Intron XP_016872443.1

View Full Product Details