Product Details

SNP ID
rs1802916
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57532077 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAATCATCTGCTGGGTGGTATCCA[A/T]GTGTGTCAGGAGCTGACCAAACTGC
Phenotype
MIM: 607376
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DCTN2 PubMed Links

Gene Details

Gene
DCTN2
Gene Name
dynactin subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261412.1 1325 Missense Mutation ATG,TTG M355L NP_001248341.1
NM_001261413.1 1325 Missense Mutation ATG,TTG M353L NP_001248342.1
NM_006400.4 1325 Missense Mutation ATG,TTG M358L NP_006391.1
XM_011537761.2 1325 Missense Mutation ATG,TTG M266L XP_011536063.1
Gene
MBD6
Gene Name
methyl-CpG binding domain protein 6
There are no transcripts associated with this gene.

View Full Product Details