Product Details

SNP ID
rs1051627
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57530237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTCTTGTAGTATAGAGTGGCTTA[A/G]ATTGCCTGTTATGACGAATGAATAT
Phenotype
MIM: 607376 MIM: 126337
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCTN2 PubMed Links

Gene Details

Gene
DCTN2
Gene Name
dynactin subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261412.1 1926 UTR 3 NP_001248341.1
NM_001261413.1 1926 UTR 3 NP_001248342.1
NM_006400.4 1926 UTR 3 NP_006391.1
XM_011537761.2 1926 UTR 3 XP_011536063.1
Gene
DDIT3
Gene Name
DNA damage inducible transcript 3
There are no transcripts associated with this gene.

Gene
MBD6
Gene Name
methyl-CpG binding domain protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052897.3 1926 Intron NP_443129.3
XM_005268617.1 1926 Intron XP_005268674.1
XM_005268618.1 1926 Intron XP_005268675.1
XM_005268620.2 1926 Intron XP_005268677.1
XM_006719217.1 1926 Intron XP_006719280.1
XM_006719218.1 1926 Intron XP_006719281.1
XM_011537851.2 1926 Intron XP_011536153.1

View Full Product Details