Product Details

SNP ID
rs1056971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50579010 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTTACATACACAGCACGGGGCT[C/T]TGGCCTGCCAGCCATGGGGACCTAC
Phenotype
MIM: 612395 MIM: 601987
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHKB PubMed Links

Gene Details

Gene
CHKB
Gene Name
choline kinase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005198.4 1577 UTR 3 NP_005189.2
Gene
CHKB-AS1
Gene Name
CHKB antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
CHKB-CPT1B
Gene Name
CHKB-CPT1B readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
CPT1B
Gene Name
carnitine palmitoyltransferase 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145134.1 1577 Intron NP_001138606.1
NM_001145135.1 1577 Intron NP_001138607.1
NM_001145137.1 1577 Intron NP_001138609.1
NM_004377.3 1577 Intron NP_004368.1
NM_152245.2 1577 Intron NP_689451.1
NM_152246.2 1577 Intron NP_689452.1

View Full Product Details