Product Details

SNP ID
rs1185771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118339630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTCAAAGTTCTAGGAGTCTTTA[G/T]GAGATTGCAAGAGTAACTCCCAGCT
Phenotype
MIM: 186790 MIM: 186740
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CD3D PubMed Links

Gene Details

Gene
CD3D
Gene Name
CD3d molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000732.4 Intron NP_000723.1
NM_001040651.1 Intron NP_001035741.1
XM_017018543.1 Intron XP_016874032.1
Gene
CD3G
Gene Name
CD3g molecule
There are no transcripts associated with this gene.

View Full Product Details